missing translation for 'onlineSavingsMsg'
Learn More

CD4 Mouse, PerCP-Cy5.5, Clone: OKT4, enQuireBio™

Mouse Monoclonal Antibody

£78.81 - £168.00

Specifications

Antigen CD4
Clone OKT4
Applications Flow Cytometry
Conjugate PerCP-Cyanine5.5
Host Species Mouse
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15951498
View Documents
EnquireBio
QAB11-PCP55-25TESTS
25 Tests
£78.81
25 tests
View alternative products
Save Up to 
»
Estimated Shipment
24-05-2021
Please to purchase this item. Need a web account? Register with us today!
15941498
View Documents
EnquireBio
QAB11-PCP55-100TESTS
100 Tests
£168.00
100 tests
View alternative products
Save Up to 
»
Estimated Shipment
24-05-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD4
Flow Cytometry
Mouse
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
920
Monoclonal
Store at 2-4°C.
OKT4
PerCP-Cyanine5.5
RUO
NP_001181945
IgG2b kappa
Protein A/G Purified
SDS