missing translation for 'onlineSavingsMsg'
Learn More

CD4 Mouse, PerCP, Clone: SK3, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB10-PCP-500Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15951478

  • £2189.43 / 500 tests
Estimated Shipment: 18-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD4
SK3
PerCP
NP_001181945
Mouse
Protein A/G Purified
RUO
Primary
920
Flow Cytometry
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD4
IgG1 kappa
500 Tests
Store at 2-4°C.
Monoclonal
Videos
SDS
Documents
Certificates
Special Offers

Special Offers