missing translation for 'onlineSavingsMsg'
Learn More

CD4 Mouse anti-Human, eFluor® 506, Clone: RPA-T4, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 69-0049-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15590637

  • £72.50 / 25 tests

In stock, ready to ship: 4

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene [RefSeq, July 2017].
Specifications

Specifications

CD4
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P01730
CD4 antigen p55, CD4 receptor, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
RPA-T4
eFluor™ 506
CD4
Liquid
CD4
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
920
Videos
SDS
Documents

For Research Use Only.