missing translation for 'onlineSavingsMsg'
Learn More

CD4, PE-Cyanine5, clone: RPA-T4, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 15-0049-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15547706

  • £162.30 / 100 tests


EU Stock 3
Add to basket

Description

Description

Description: The RPA-T4 monoclonal antibody reacts with human CD4, a 59kDa cell surface receptor expressed by a majority of thymocytes, subpopulation of mature T cells (T-helper cells) and in low levels on monocytes. CD4 is a receptor for the human immunodeficiency virus (HIV). RPA-T4 blocks HIV binding and mixed lymphocyte reaction. The RPA-T4 antibody recognizes a different epitope than the OKT4 monoclonal antibody, and these antibodies do not cross-block binding to each other's respective epitopes.Applications Reported: The RPA-T4 antibody has been reported for use in flow cytometric analysis.Applications Tested: This RPA-T4 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 667 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD4
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P01730
CD4 antigen p55, CD4 receptor, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
RPA-T4
PE-Cyanine5
CD4
Conjugated
CD4
IgG1, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
920
Videos

For Research Use Only.