CD4 Mouse anti-Human, eVolve 655, Clone: SK3 (SK-3), eBioscience™

Mouse Monoclonal Antibody

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: 86-0047-41

Code: NEW

Additional Details:
Additional Details: Weight: 0.23750kg



Disclaimers: For Research Use Only.

Product Code. 15510827

Quantity Price
1 £ 110.05 / 25 tests
Estimated Shipment date
from Supplier 06-06-2017
Add to basket


Description and Specification

Specification

Antigen CD4
Applications Flow Cytometry
Clone SK3 (SK-3)
Concentration 5μL
Conjugate eVolve 655
Format Conjugated
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Gene Alias Leu-3, T4
Host Species Murine
Isotype IgG1
Quantity 25 Tests
Regulatory Status RUO
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This product should be used within 6 months from the date of receipt.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

The SK3 monoclonal antibody reacts with human CD4, a 59kDa cell surface receptor expressed by a majority of thymocytes, a subpopulation of mature T helper cells, and at low levels on monocytes. CD4 is a receptor for the human immunodeficiency virus (HIV). SK3 blocks HIV binding and mixed lymphocyte reaction. The SK3 and RPA-T4 monoclonal antibodies do not cross-block binding, suggesting recognition of distinct epitopes.