missing translation for 'onlineSavingsMsg'
Learn More

CD4, eFluor 450, clone: SK3 (SK-3), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-0047-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15599616

  • £171.42 / 100 tests


EU Stock 10
Add to basket

Description

Description

Description: The SK3 monoclonal antibody reacts with human CD4, a 59-kDa cell surface receptor expressed by a majority of thymocytes, a subpopulation of mature T helper cells, and, in low levels, monocytes. CD4 is a receptor for the human immunodeficiency virus (HIV). SK3 blocks HIV binding and mixed lymphocyte reaction. The SK3 and RPA-T4 monoclonal antibodies do not cross-block binding, suggesting recognition of distinct epitopes.Applications Reported: This SK3 (SK-3) antibody has been reported for use in flow cytometric analysis.Applications Tested: This SK3 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD4
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P01730
CD4 antigen p55, CD4 receptor, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
SK3 (SK-3)
eFluor 450
CD4
Conjugated
CD4
IgG1, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
920
Videos

For Research Use Only.