missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Goat anti-Feline, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | CD4 |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant feline CD4 |
Gene ID (Entrez) | 920 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Reconstitution | Reconstitute at 0.2 mg/mL in sterile PBS. |
Description
CD4 Polyclonal specifically detects CD4 in Feline samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
CD4 | |
920 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Goat | |
IgG | |
Affinity Purified | |
RUO |
Mouse myeloma cell line NS0-derived recombinant feline CD4 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects feline CD4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant canine CD4 is observed and 5% cross-reactivity with recombinant human CD4 and recombinant mouse CD4 is observed. | |
Unconjugated | |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only