Learn More
Epredia™ Lab Vision™ CD4 Ab-8, Mouse Monoclonal Antibody
Obtain accurate and reproducible results in immunohistochemistry procedures with Epredia™ CD4 Ab-8, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1528-R7
Code : LD
Additional Details : Weight : 0.02700kg
Description
CD4, a single chain transmembrane glycoprotein, is found on a T cell subset (helper/inducer) representing 45% of peripheral blood lymphocytes. It is also present on 80% of thymocytes and at a lower level on monocytes. It is involved in recognition of antigen presented along with MHC class II by APCs. It serves as receptor for HIV.Host Species: Mouse
Clone: 4B12
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: Stimulated human leukocytes
Molecular Weight: 59 kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
CD4 Ab-8 | |
Monoclonal | |
Unconjugated | |
Stimulated human leukocytes | |
IVD | |
Primary | |
Prediluted |
Immunohistochemistry (Paraffin) | |
4B12 | |
Mouse | |
7 mL | |
T-Cell | |
Human | |
IgG2a κ |