missing translation for 'onlineSavingsMsg'
Learn More

CD3e, FITC, clone: eBio500A2 (500A2), eBioscience™

Syrian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0033-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15207989

  • £37.33 / 50µg


Estimated Shipment date
from Supplier 29-06-2018
Add to basket

Description

Description

Description: The eBio500A2 monoclonal antibody reacts with the 25 kD epsilon subunit of the mouse CD3 complex. CD3 subunits gamma, delta and ε are required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells, as well as NKT cells. Binding of eBio500A2 to CD3 initiates the intracellular biochemical pathway resulting in cellular activation and proliferation. The 500A2 antibody is able to partially cross-block the 17A2 and 145-2C11 antibodies, indicating that all three of these anti-CD3 antibodies recognize distinct but overlapping epitopes.Applications Reported: This eBio500A2 (500A2) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBio500A2 (500A2) antibody has been tested by flow cytometric analysis. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD3e
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P22646
FLJ18683, T3E, TCRE, CD3-epsilon, CD3e antigen, epsilon polypeptide TiT3 complex, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain
Golden Syrian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
eBio500A2 (500A2)
FITC
Cd3e
Conjugated
Cd3e
IgG
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
12501
Videos

For Research Use Only.