missing translation for 'onlineSavingsMsg'
Learn More

CD3e, eFluor 450, clone: eBio500A2 (500A2), eBioscience™

Syrian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-0033-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15238867

  • £73.79 / 25µg


EU Stock 1
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The eBio500A2 monoclonal antibody reacts with the 25 kD epsilon subunit of the mouse CD3 complex. CD3 subunits gamma, delta and ε are required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells, as well as NKT cells. Binding of eBio500A2 to CD3 initiates the intracellular biochemical pathway resulting in cellular activation and proliferation. The 500A2 antibody is able to partially cross-block the 17A2 and 145-2C11 antibodies, indicating that all three of these anti-CD3 antibodies recognize distinct but overlapping epitopes.Applications Reported: This eBio500A2 (500A2) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBio500A2 (500A2) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD3e
eBio500A2 (500A2)
eFluor 450
Conjugated
Golden Syrian Hamster
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
CD3 epsilon
IgG
RUO
Primary
Murine
Videos