Learn More
CD3e Armenian Hamster, FITC, Clone: 145-2C11, enQuireBio™
Armenian Hamster Monoclonal Antibody
Brand:  EnquireBio QAB2-F-500ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15951638
  • £164.00 / 500µg
Estimated Shipment: 12-06-2024
to see stock.
Specifications
CD3e
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Cd3e
Protein A/G Purified
RUO
12501
IgG
Flow Cytometry
145-2C11
FITC
XP_006510029
Armenian Hamster
500 μg
Primary
Store at 2-4°C.