missing translation for 'onlineSavingsMsg'
Learn More

CD3e, FITC, clone: 145-2C11, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0031-63

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15257979

  • £24.69 / 10Āµg


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Description

Description

Description: The 145-2C11 monoclonal antibody reacts with mouse CD3e, a 20kDa subunit of the TCR complex. Along with the other CD3 subunits, gamma and delta, the epsilon chain is required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. Binding of 145-2C11 to TCR initiates the intracellular biochemical pathway resulting in cellular activation, proliferation, and apoptosis depending on specific conditions utilized. 145-2C11 is commonly used as a phenotypic marker for mouse T cells.Applications Reported: This 145-2C11 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining of frozen tissue sections.Applications Tested: The 145-2C11 antibody has been tested by flow cytometric analysis of mouse thymocytes and splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. This 145-2C11 antibody has been tested by immunohistochemistry on frozen mouse spleen and can be used at less than or equal to 10 ug/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD3e
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P22646
FLJ18683, T3E, TCRE, CD3-epsilon, CD3e antigen, epsilon polypeptide TiT3 complex, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow CytometryĆ¾Immunohistochemistry (Frozen)
145-2C11
FITC
Cd3e
Conjugated
Cd3e
IgG
10μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
12501
Videos

For Research Use Only.