Learn More
CD39 Monoclonal Antibody (24DMS1), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£151.00 - £285.00
Specifications
Antigen | CD39 |
---|---|
Clone | 24DMS1 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15320920
|
Affymetrix eBioscience
25-0391-80 |
25 μg |
£151.00
25µg |
Estimated Shipment: 01-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15330920
|
Affymetrix eBioscience
25-0391-82 |
100 μg |
£285.00
100µg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The 24DMS1 monoclonal antibody reacts with mouse CD39, also known as NTPDaseI. E-NTPDases are enzymes that convert nucleoside tri- and diphosphates (NTDPs) into nucleoside monophosphate (NMP), thereby removing toxic extracellular ATP and ADP. CD39 is the domit member of this family in the immune system and is involved in suppression of inflammation and control of platelet activation. CD39 can impact expression of CD73, another E-NTPase. Together, these molecules influence inflammation responses. CD39 is expressed on B cells, Langerhans cells and most monocytes. In addition, CD39 is found on a subset of CD4+ T cells that are mostly CD25+FoxP3+ T reg cells. T reg cells from CD39-null mice showed impaired suppressive properties in vitro and in vivo. Applications Reported: This 24DMS1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 24DMS1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive photo-induced oxidation.
CD39 Monoclonal antibody specifically detects CD39 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD39 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
2610206B08Rik; AA408691; ATPDase; Cd39; CD39 antigen; DKFZp686D194; DKFZp686I093; ecto-apyrase; Ecto-ATP diphosphohydrolase 1; ecto-ATPase 1; ecto-ATPDase 1; Ectonucleoside triphosphate diphosphohydrolase 1; ENTPD1; FLJ40921; FLJ40959; Lymphoid cell activation antigen; NTPDase 1; NTPDase-1; SPG64 | |
ENTPD1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
ENTPD1 |
24DMS1 | |
Flow Cytometry | |
PE-Cyanine7 | |
Rat | |
Mouse | |
P55772 | |
12495 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.