missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD39 Monoclonal Antibody (A1), Alexa Fluor™ 488, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD3920
Code : 50
Additional Details : Weight : 0.50000kg
Description
Based on our testing, publications, and results reported from customers, the Alexa Fluor® 488 dye provides the best fluorescein (FITC) substitute.
CD39 Monoclonal antibody specifically detects CD39 in Human samples. It is validated for Flow Cytometry.Specifications
CD39 | |
Monoclonal | |
Alexa Fluor 488 | |
ENTPD1 | |
2610206B08Rik; AA408691; ATPDase; Cd39; CD39 antigen; DKFZp686D194; DKFZp686I093; ecto-apyrase; Ecto-ATP diphosphohydrolase 1; ecto-ATPase 1; ecto-ATPDase 1; Ectonucleoside triphosphate diphosphohydrolase 1; ENTPD1; FLJ40921; FLJ40959; Lymphoid cell activation antigen; NTPDase 1; NTPDase-1; SPG64 | |
Mouse | |
purified | |
ASR | |
953 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
A1 | |
PBS with 4mg/mL BSA and 0.1% sodium azide | |
P49961 | |
ENTPD1 | |
Human CD39 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |