Learn More
CD38 Monoclonal Antibody (HB7), Super Bright™ 702, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£147.00 - £319.00
Specifications
Antigen | CD38 |
---|---|
Clone | HB7 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15875723
|
Affymetrix eBioscience
67-0388-42 |
100 Tests |
£319.00
Pack of 100 |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15865723
|
Affymetrix eBioscience
67-0388-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The HIT2 monoclonal antibody reacts with the human CD38 molecule, an approximately 45 kDa type II transmembrane protein. CD38 is expressed by thymocytes, peripheral B cells including plasma cells, activated T cells, and monocytes. CD38 is a counter-receptor for CD31 and has both cyclase and hydrolase enzymatic activity. Applications Reported: This HIT2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HIT2 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 702 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 702 nm. We recommend using a 710/50 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No. SB-4400-42) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation.
CD38 Monoclonal antibody specifically detects CD38 in Human samples. It is validated for Flow Cytometry.Specifications
CD38 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P28907 | |
CD38 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD38 |
HB7 | |
Flow Cytometry | |
Super Bright 702 | |
Mouse | |
Human | |
952 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |