missing translation for 'onlineSavingsMsg'
Learn More

CD38, clone: 90, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0381-82

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15216927

  • £68.51 / 100µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The 90 monoclonal antibody reacts with the mouse CD38 molecule, an ∽42kDa type II transmembrane protein. CD38 is expressed at increasingly higher levels on B cells at each stage of B-cell differentiation, and is then down-regulated on germinal center B cells and mature plasma cells. Its expression is reported on a subpopulation of thymocytes, mature T cells, and NK cells. Crosslinking of CD38 on the surface of mature, resting B cells induces B-cell proliferation, which is enhanced by co-signals such as IL-4 and LPS. CD38, a counter-receptor for CD31, is an ectoenzyme with cyclase and hydrolase enzymatic activity and is speculated to play a role in lymphocyte activation and differentiation.Applications Reported: The 90 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining. It has also been reported in functional assays. (Please use Functional Grade purified 90, cat. 16-0381, in functional assays.).Applications Tested: The 90 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD38
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P56528
T10, ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, CD38 antigen p45, NAD+ nucleosidase, cADPr hydrolase 1, cyclic ADP-ribose hydrolase 2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow CytometryþFunctional AssayþImmunohistochemistry
90
Unconjugated
Cd38
Purified
Cd38
IgG2a, kappa
100μg
Store at 2-8°C.
Primary
12494
Videos

For Research Use Only.