Learn More
CD371 (Clec12A) Monoclonal Antibody (HB3), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£90.00 - £224.00
Specifications
Antigen | CD371 (Clec12A) |
---|---|
Clone | HB3 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15507266
|
Affymetrix eBioscience
12-9878-41 |
25 Tests |
£90.00
25 tests |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15517266
|
Affymetrix eBioscience
12-9878-42 |
100 Tests |
£224.00
100 tests |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: This HB3 monoclonal antibody reacts with human CD371, which is also known as Clec12A or Myeloid Inhibitory C-type Lectin-like receptor (MICL). CD371 is a highly glycosylated transmembrane receptor with a cytoplasmic ITIM that associates with SHP1 and SHP2. CD371 is expressed mainly on granulocytes, monocytes, macrophages, and dendritic cells, including plasmacytoid dendritic cells. Expression of CD371 on NK cells remains controversial. Although the function of CD371 is not fully understood, studies suggest that this receptor plays a role in inflammation and LPS activation. Applications Reported: This HB3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HB3 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD371 is also known as Clec12A or Myeloid Inhibitory C-type Lectin-like receptor (MICL). CD371 is a highly glycosylated transmembrane receptor with a cytoplasmic ITIM that associates with SHP1 and SHP2. CD371 is expressed mainly on granulocytes, monocytes, macrophages, and dendritic cells, including plasmacytoid dendritic cells. Expression of CD371 on NK cells remains controversial. Although the function of CD371 is not fully understood, studies suggest that this receptor plays a role in inflammation and LPS activation.Specifications
CD371 (Clec12A) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
CD371; Clec12a; CLL1; CLL-1; C-type lectin domain family 12 member A; C-type lectin domain family 12, member A; C-type lectin protein CLL-1; C-type lectin-like molecule 1; C-type lectin-like molecule-1; D230024O04; DCAL2; DCAL-2; Dendritic cell-associated lectin 2; KLRL1; Micl; Myeloid inhibitory C-type lectin-like receptor | |
CLEC12A | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CLEC12A |
HB3 | |
Flow Cytometry | |
PE | |
Mouse | |
Human | |
Q5QGZ9 | |
160364 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.