missing translation for 'onlineSavingsMsg'
Learn More

CD370 (Clec9A), PerCP-eFluor™ 710, clone: 42D2, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-5975-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15341380

  • £85.50 / 25µg


Estimated Shipment
29-01-2019
Add to basket

Description

Description

Description: This 42D2 monoclonal antibody reacts with mouse CD370, which is also known as DNGR-1 or Clec9A. CD370 is a C-type lectin-like receptor expressed on conventional and plasmacytoid dendritic cells (DCs). In particular, CD8+ conventional DCs in the thymus, spleen, and lymph nodes, as well as CD24+ DCs in the blood, express CD370. This receptor is not detectable on most B cells, T and NK cells, monocytes, and macrophages. The cytoplasmic domain of CD370 contains an ITAM motif that has been demonstrated to phosphorylate Syk tyrosine kinase, leading to NFAT activation. This signaling reportedly mediates the presentation of necrotic cell-associated antigens by CD8+ DCs in innate immunity.Applications Reported: This 42D2 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 42D2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.PerCP-eFluor™ 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative. Our testing indicates that PerCP-eFluor™ 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4°C, but please evaluate for alternative fixation protocols.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD370 (Clec9A)
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q8BRU4
UNQ9341/PRO34046, DNGR1, UNQ9341, C-type lectin domain family 9 member A, HEEE9341
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
42D2
PerCP-eFluor 710
Clec9a
Liquid
Clec9a
IgG1
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
232414
Videos

For Research Use Only.