missing translation for 'onlineSavingsMsg'
Learn More

CD369 (Clec7A, Dectin-1) Mouse anti-Human, Non-human primate, PerCP-eFluor 710, Clone: 15E2, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-9856-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15559446

  • £220.84 / 100 tests


EU Stock 4
Add to basket

Disclaimers:

For Research Use Only.


Specifications

Description

This 15E2 monoclonal antibody reacts with human and non-human primate CD369, which is also known as the Dectin-1 or beta-glucan receptor or CLEC7A. CD369 is expressed predominantly on monocytes, macrophages, and dendritic cells. Moreover, this receptor can also be detected on neutrophils and lymphocytes, as well as induced by cytokines such as IL-4 or LPS. In humans, CD369 exists as two isoforms that arise due to alternative splicing. CD369 binds beta-1,3-glucan and plays an important role in activating signal transduction involved in the innate immune response to fungal and mycobacterial infections. Studies have also shown the involvement of CD369 in dendritic cell activation.

The 15E2 monoclonal antibody has been reported to recognize both isoforms of CD369 and activate dendritic cells. It does not crossreact to mouse CD369.
Description & Specifications

Specifications

CD369 (Clec7A, Dectin-1)
1.50E+03
PerCP-eFluor 710
Conjugated
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Beta-glucan receptor, ClecSF12
IgG2a
RUO
Primary
Non-human Primate