Learn More
CD369 (Clec7a, Dectin-1) Monoclonal Antibody (15E2), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£156.00 - £262.00
Specifications
Antigen | CD369 (Clec7a, Dectin-1) |
---|---|
Clone | 15E2 |
Concentration | 1 mg/mL |
Applications | Functional Assay |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15527836
|
Affymetrix eBioscience
16-9856-82 |
100 μg |
£156.00
100µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15537836
|
Affymetrix eBioscience
16-9856-85 |
500 μg |
£262.00
500µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: This 15E2 monoclonal antibody reacts with human and non-human primate CD369, which is also known as the Dectin-1 or beta-glucan receptor or CLEC7A. CD369 is expressed predomitly on monocytes, macrophages, and dendritic cells. Moreover, this receptor can also be detected on neutrophils and lymphocytes, as well as induced by cytokines such as IL-4 or LPS. In humans, CD369 exists as two isoforms that arise due to alternative splicing. CD369 binds beta-1,3-glucan and plays an important role in activating signal transduction involved in the innate immune response to fungal and mycobacterial infections. Studies have also shown the involvement of CD369 in dendritic cell activation. The 15E2 monoclonal antibody has been reported to recognize both isoforms of CD369 and activate dendritic cells. It does not crossreact to mouse CD369. Applications Reported: The 15E2 antibody has been reported to activate dendritic cells. Applications Tested: This 15E2 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Endotoxin: Less than 0.
CLEC7A, also known as dectin-1, is a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily and is predominantly expressed on myeloid cells. It is a small glycoprotein type II membrane receptor with an extracellular C-type lectin-like domain fold and a cytoplasmic domain with an immunoreceptor tyrosine-based activation motif (ITAM). CLEC7A functions as a pattern-recognition receptor that recognizes a variety of beta-1,3-linked and beta-1,6-linked glucans from fungi and plants, and in this way plays a role in innate immune response. Upon fungal exposure, CLEC7A activates Syk tyrosine kinase, triggering a massive oxidative burst through the formation of reactive oxygen species.Specifications
CD369 (Clec7a, Dectin-1) | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.2 | |
Beta-glucan receptor; beta-GR; BGR; CANDF4; CD369; CLEC7A; Clec7b; CLECSF12; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 12; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 12; C-type lectin domain family 7 member A; C-type lectin domain family 7, member A; C-type lectin superfamily member 12; DC-associated C-type lectin 1; DECTIN1; dectin-1; Dendritic cell-associated C-type lectin 1; dendritic cell-associated C-type lectin-1; lectin-like receptor 1; RGD1565140; SCARE2; UNQ539/PRO1082 | |
Clec7a | |
Primary | |
4° C | |
Clec7a |
15E2 | |
Functional Assay | |
Functional Grade | |
Mouse | |
Human, Non-human Primate | |
Q9BXN2 | |
64581 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.