missing translation for 'onlineSavingsMsg'
Learn More

CD369 (Clec7a, Dectin-1) Mouse anti-Human, Non-human primate, Functional Grade, Clone: 15E2, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 16-9856-85

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15537836

  • £216.00 / 500µg

Estimated Shipment: 02-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: This 15E2 monoclonal antibody reacts with human and non-human primate CD369, which is also known as the Dectin-1 or beta-glucan receptor or CLEC7A. CD369 is expressed predomitly on monocytes, macrophages, and dendritic cells. Moreover, this receptor can also be detected on neutrophils and lymphocytes, as well as induced by cytokines such as IL-4 or LPS. In humans, CD369 exists as two isoforms that arise due to alternative splicing. CD369 binds beta-1,3-glucan and plays an important role in activating signal transduction involved in the innate immune response to fungal and mycobacterial infections. Studies have also shown the involvement of CD369 in dendritic cell activation. The 15E2 monoclonal antibody has been reported to recognize both isoforms of CD369 and activate dendritic cells. It does not crossreact to mouse CD369. Applications Reported: The 15E2 antibody has been reported to activate dendritic cells. Applications Tested: This 15E2 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Endotoxin: Less than 0.

001 ng/ug antibody as determined by the LAL assay. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC. CLEC7A, also known as dectin-1, is a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily and is predomitly expressed on myeloid cells. It is a small glycoprotein type II membrane receptor with an extracellular C-type lectin-like domain fold and a cytoplasmic domain with an immunoreceptor tyrosine-based activation motif (ITAM). CLEC7A functions as a pattern-recognition receptor that recognizes a variety of beta-1,3-linked and beta-1,6-linked glucans from fungi and plants, and in this way plays a role in innate immune response. Upon fungal exposure, CLEC7A activates Syk tyrosine kinase, triggering a massive oxidative burst through the formation of reactive oxygen species.
Specifications

Specifications

CD369 (Clec7A, Dectin-1)
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.2
Q9BXN2
UNQ539/PRO1082, BGR, CANDF4, CLECSF12, DECTIN1,C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 12; C-type lectin domain family 7 member A; C-type lectin superfamily member 12; DC-associated C-type lectin 1; beta-glucan receptor; dectin-1; dendritic cell-associated C-type lectin 1; dendritic cell-associated C-type lectin-1; lectin-like receptor 1
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Non-human Primate
Functional Assay
15E2
Functional Grade
CLEC7A
Liquid
CLEC7A
IgG2a, kappa
500 μg
4° C
Primary
64581
Videos
SDS
Documents
Certificates

For Research Use Only.