Learn More
CD368 (Clec6) Monoclonal Antibody (9B9), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 50-9113-41
71.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.23750kg
Description
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.
CD368 (Clec6) Monoclonal antibody specifically detects CD368 (Clec6) in Human samples. It is validated for Flow Cytometry.Specifications
CD368 (Clec6) | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Q8WXI8 | |
CLEC4D | |
Affinity chromatography | |
RUO | |
338339 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
9B9 | |
eFluor 660 | |
CLEC4D | |
CD368; Clec4d; CLEC6; CLEC-6; CLECSF8; CLECSF8Mpcl; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 8; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 8; C-type lectin domain family 4 member D; C-type lectin domain family 4, member d; C-type lectin receptor; C-type lectin superfamily member 8; C-type lectin, superfamily member 8; C-type lectin-like receptor 6; macrophage C-type lectin; macrophage-restricted C-type lectin; Mcl; MGC40078; Mpcl; superfamilymember 8 | |
Mouse | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
For Research Use Only.