missing translation for 'onlineSavingsMsg'
Learn More

CD360 (IL-21 Receptor), PE, clone: 2SX21R, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-3601-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15516976

  • £74.42 / 25 tests


Estimated Shipment date
from Supplier 28-06-2018
Add to basket

Description

Description

Description: This 2SX21R monoclonal antibody reacts with human CD360 (Interleukin-21 Receptor, IL-21R). IL-21R forms a heterodimeric receptor complex with the common gamma chain to form a functional receptor for IL-21. IL-21 is a class I cytokine of the common gamma chain family of cytokines and signals through the IL-21R complex to activate the JAK/STAT (Janus Kinase/Signal Transducers and Activators of Transcription) signaling pathway. Activation of the JAK/STAT pathway by IL-21 promotes transcription of genes responsive to STAT1 and STAT3. IL-21 is mainly produced by CD4+ T cells and mediates the proliferation, differentiation, and effector functions of cells expressing IL-21R such as B cells, T cells, NK cells, dendritic cells and other populations of myeloid cells. IL-21 and its receptor are implicated in the pathogenesis of chronic viral infections, autoimmunity, allergy, organ transplantation, and cancer.Applications Reported: This 2SX21R antibody has been reported for use in flow cytometric analysis.Applications Tested: This 2SX21R antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD360 (IL-21 Receptor)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q9HBE5
Interleukin-21 Receptor
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
2SX21R
PE
IL21R
Conjugated
IL21R
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
50615
Videos

For Research Use Only.