missing translation for 'onlineSavingsMsg'
Learn More

CD36, clone: eBioNL07 (NL07), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0369-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15286917

  • £37.00 / 25µg


EU Stock 1
Add to basket

Description

Description

Description: The monoclonal antibody eBioNL07 recognizes human CD36, which is a member of the class B scavenger receptor family. CD36 was originally identified as a platelet-membrane glycoprotein also called glycoprotein IV and a receptor for thrombospondin-1 (TSP-1) and extracellular matrix proteins. Binding to TSP-1 is in the CLESH (CD36 LIMP-II Emp sequence homology) domain of CD36. CD36 expression is broad and includes microvascular (but not large vessel) endothelium, adipocytes, skeletal muscle, dendritic cells, epithelia of the retina, breast, and intestine, smooth muscle cells, and hematopoietic cells, including erythroid precursors, platelets, monocytes/macrophages, DCs and megakaryocytes. Expression on platelets is absent on Nak^a negative donors. Unlike other scavenger receptor, CD36 binds LDL that has been exposed to minimally oxidizing conditions. CD36 is also a fatty acid translocase (FAT) necessary for the transport of long-chain fatty acids (LCFAs) and therefore may play a role in atherosclerosis.Applications Reported: This eBioNL07 (NL07) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBioNL07 (NL07) antibody has been tested by flow cytometric analysis of human peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD36
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P16671
FAT, GPIV, Scarb3, GPIIIB, PAS IV, PAS-4, fatty acid translocase, glycoprotein IIIb, platelet glycoprotein 4, platelet glycoprotein IV
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
eBioNL07 (NL07)
Unconjugated
CD36
Purified
CD36
IgM
25μg
Store at 2-8°C.
Primary
948
Videos

For Research Use Only.