Learn More
CD357 / GITR Rat, FITC, Clone: DTA-1, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB72-F-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15962408
  • £220.56 / 100µg
Estimated Shipment: 17-01-2022
Specifications
CD357 / GITR
DTA-1
FITC
NP_033426
Rat
Protein A/G Purified
RUO
Primary
21936
Flow Cytometry
0.5mg/mL
10mM Tris, 150 mM NaCl, 0.05% BSA, 0.05% NaN3, pH8
Tnfrsf18
IgG2b
100 μg
Store at 2-4°C.