missing translation for 'onlineSavingsMsg'
Learn More

CD357 (AITR/GITR), PE, clone: DTA-1, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5874-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15208869

  • £74.85 / 25µg


Estimated Shipment date
from Supplier 30-05-2018
Add to basket

Description

Description

Description: The DTA-1 monoclonal antibody reacts with mouse GITR, Glucocorticoid-Induced TNFR family gene, also known as TNFRSF18. In naive mice, GITR is expressed predominantly by CD4+CD25+ T regulatory cells (Treg) and by CD25+ CD4+ CD8- thymocytes. Stimulation of GITR with DTA-1 antibody abrogates Treg cell-mediated suppression. Although it does not deplete CD357 expressing cells in vivo, it has been shown to induce tumorogenesis. The removal of GITR-expressing Treg cells or the administration of DTA-1 resulted in organ specific autoimmune disease.Applications Reported: The DTA-1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This DTA-1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.004 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD357
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
O35714
AITR, GITR-D, TNFRSF18
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
DTA-1
PE
Tnfrsf18
Conjugated
Tnfrsf18
IgG2b, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
21936
Videos

For Research Use Only.