missing translation for 'onlineSavingsMsg'
Learn More

CD357 (AITR/GITR), FITC, clone: DTA-1, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-5874-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15278269

  • £68.96 / 25µg


EU Stock 1
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The DTA-1 monoclonal antibody reacts with mouse GITR, Glucocorticoid-Induced TNFR family gene, also known as TNFRSF18. In naive mice, GITR is expressed predominantly by CD4+CD25+ T regulatory cells (Treg) and by CD25+ CD4+ CD8- thymocytes. Stimulation of GITR with DTA-1 antibody abrogates Treg cell-mediated suppression. Although it does not deplete CD357 expressing cells in vivo, it has been shown to induce tumorogenesis. The removal of GITR-expressing Treg cells or the administration of DTA-1 resulted in organ specific autoimmune disease.Applications Reported: The DTA-1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This DTA-1 antibody has been tested flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD357
DTA-1
FITC
Conjugated
Rat
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TNFRSF18, AITR, glucocorticoid-induced TNFR-related
IgG2b
RUO
Primary
Murine
Videos