Learn More
CD357 (AITR/GITR) Monoclonal Antibody (DTA-1), eFluor™ 450, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£100.00 - £237.00
Specifications
Antigen | CD357 (AITR/GITR) |
---|---|
Clone | DTA-1 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15301800
|
Affymetrix eBioscience
48-5874-82 |
100 μg |
£237.00
100µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15391790
|
Affymetrix eBioscience
48-5874-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The DTA-1 monoclonal antibody reacts with mouse GITR, Glucocorticoid-Induced TNFR family gene, also known as TNFRSF18. In naive mice, GITR is expressed predominantly by CD4+CD25+ T regulatory cells (Treg) and by CD25+ CD4+ CD8- thymocytes. Stimulation of GITR with DTA-1 antibody abrogates Treg cell-mediated suppression. Although it does not deplete CD357 expressing cells in vivo, it has been shown to induce tumorogenesis. The removal of GITR-expressing Treg cells or the administration of DTA-1 resulted in organ specific autoimmune disease. Applications Reported: This DTA-1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This DTA-1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 450 is an alternative to Pacific Blue™. eFluor™ 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD357 (AITR/GITR) Monoclonal antibody specifically detects CD357 (AITR/GITR) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD357 (AITR/GITR) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
O35714 | |
Tnfrsf18 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Tnfrsf18 |
DTA-1 | |
Flow Cytometry | |
eFluor 450 | |
Rat | |
Mouse | |
21936 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.