Learn More
CD357 (AITR/GITR) Monoclonal Antibody (DTA-1), Super Bright™ 600, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£310.00
Specifications
Antigen | CD357 (AITR/GITR) |
---|---|
Clone | DTA-1 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15913564
|
Affymetrix eBioscience
63-5874-82 |
100 μg |
£310.00
100µg |
Estimated Shipment: 15-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The DTA-1 monoclonal antibody reacts with mouse GITR, Glucocorticoid-Induced TNFR family gene, also known as TNFRSF18. In naive mice, GITR is expressed predomitly by CD4+CD25+ T regulatory cells (Treg) and by CD25+ CD4+ CD8- thymocytes. Stimulation of GITR with DTA-1 antibody abrogates Treg cell-mediated suppression. Although it does not deplete CD357 expressing cells in vivo, it has been shown to induce tumorogenesis. The removal of GITR-expressing Treg cells or the administration of DTA-1 resulted in organ specific autoimmune disease. Applications Reported: This DTA-1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This DTA-1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.03 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 600 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 600 nm. We recommend using a 610/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome.
CD357 (AITR/GITR) Monoclonal antibody specifically detects CD357 (AITR/GITR) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD357 (AITR/GITR) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
O35714 | |
Tnfrsf18 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Tnfrsf18 |
DTA-1 | |
Flow Cytometry | |
Super Bright 600 | |
Rat | |
Mouse | |
21936 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |