missing translation for 'onlineSavingsMsg'
Learn More

CD34 Mouse anti-Human, PE-eFluor® 610, Clone: 4H11, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 61-0349-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15590427

  • £200.00 / 100 tests

Estimated Shipment
08-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PE-eFluor™ 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red™). Please make sure that your instrument is capable of detecting this fluorochome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. CD34 is a highly glycosylated, monomeric, 111-115 kDa surface protein that is present on many stem cell populations. CD34 is a stem cell marker although its expression on human hematopoietic stem cells is reversible. CD34 may serve as a surface receptor that undergoes receptor-mediated endocytosis and regulates adhesion, differentiation, and proliferation of hematopoietic stem cells and other progenitors. CD34 expression is likely to represent a specific state of hematopoietic development that may have altered adhering properties with expanding and differentiating capabilities in both in vitro and in vivo conditions. CD34 is possibly an adhesion molecule with a putative role for mediating the attachment of stem cells to the bone marrow extracellular matrix or directly to stromal cells. Further, CD34 could act as a scaffold for the attachment of lineage specific glycans, allowing stem cells to bind to lectins expressed by stromal cells or other marrow components. CD34 is thought to have a role in presenting carbohydrate ligands to selectins. The intracellular chain of the CD34 antigen is a site of phosphorylation by activated protein kinase C suggesting a putative role in signal transduction. Diseases associated with CD34 dysfunction include dermatofibrosarcoma and neurofibroma.
Specifications

Specifications

CD34
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P28906
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
4H11
PE-eFluor™ 610
CD34
Liquid
CD34
IgG1, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
947
Videos
Documents

For Research Use Only.