missing translation for 'onlineSavingsMsg'
Learn More

CD34, Biotin, clone: 4H11, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0349-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15289529

  • £62.63 / 25µg


Estimated Shipment date
from Supplier 29-06-2018
Add to basket

Description

Description

Description: The 4H11 monoclonal antibody reacts with human CD34, also known as mucosialin. CD34 belongs to a protein family which also includes endoglycan and podocalyxin. Members of this family are single pass transmembrane proteins with a heavily glycosylated extracellular and N-terminal mucin domain. CD34 was first identified as an antigen expressed on hematopoietic progenitors, and has since been extensively used as a marker to isolate cells capable of hematopoietic cell engraftment. In spite of this, the function of CD34 remains unresolved. In addition to expression on hematopoietic progenitors, CD34 is expressed on some populations of mesenchymal stem cells, tumor cell lines, and by vascular endothelia in the adult. Epitopes of CD34 have been assigned to three classes (class I, II or III) based on their differential sensitivity to enzymatic cleavage by neuraminidase, chymopapain, or O-glycoprotease. According to this analysis, the 4H11 antibody belongs to class III, indicating that it reacts with a protein epitope.Applications Reported: This 4H11 (APG) antibody has been reported for use in flow cytometric analysis.Applications Tested: This 4H11 (APG) antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD34
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P28906
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
4H11
Biotin
CD34
Conjugated
CD34
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze.
Primary
947
Videos

For Research Use Only.