missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD34 (Clone QBEnd/10) Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry procedures with Epredia™ CD34 (Endothelial Cell Marker) Ab-1, Mouse Monoclonal Antibody.
£111.00 - £685.00
Specifications
Antigen | CD34 (Endothelial Cell Marker) Ab-1 |
---|---|
Clone | QBEnd/10 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Host Species | Mouse |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
15230675
|
Epredia
MS-363-P0 |
Concentrated | 100 μL |
£111.00
100µL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15220675
|
Epredia
MS-363-P |
Concentrated | 1 mL |
£685.00
1mL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15240675
|
Epredia
MS-363-R7 |
Prediluted | 7 mL |
£215.00
7mL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
CD34, a single chain transmembrane glycoprotein, is selectively expressed on human lymphoid and myeloid hematopoietic progenitor cells. Staining for CD34 has been used to measure angiogenesis, which reportedly predicts tumor recurrence. Antibody to CD34 also reacts with vascular endothelial cells in normal tissues and in benign and malignant proliferations. This antibody is of value in the study of benign and malignant vascular tumors as well as characterization of leukemias.Host Species: Mouse
Clone: QBEnd/10
Isotype: IgG1
Species Reactivity: Human. Does not react with dog and rat. Others not known
Immunogen: Detergent solubilized vesicular suspension prepared from a perfusate of human term placenta
Molecular Weight: 105-120 kDa
Positive Control: Tonsil
Cellular Localization: Membrane/cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
CD34 (Endothelial Cell Marker) Ab-1 | |
Immunohistochemistry (Paraffin) | |
Mouse | |
Oncogenesis | |
Detergent solubilized vesicular suspension prepared from a perfusate of human term placenta | |
Primary |
QBEnd/10 | |
Monoclonal | |
IVD | |
Human | |
IgG1 |