missing translation for 'onlineSavingsMsg'
Learn More

CD335 (NKp46) Rat anti-Mouse, PE-eFluor® 610, Clone: 29A1.4, eBioscience™

Rat Monoclonal Antibody

£81.00 - £187.00

Specifications

Antigen CD335 (NKp46)
Clone 29A1.4
Target Species Mouse
Conjugate PE-eFluor™ 610
Isotype IgG2a, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15560487 View Documents Affymetrix eBioscience
61-3351-80
25 μg £81.00
25µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 14-05-2020
Please to purchase this item. Need a web account? Register with us today!
15570487 View Documents Affymetrix eBioscience
61-3351-82
100 μg £187.00
100µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 16-04-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PE-eFluor® 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red®). Please make sure that your instrument is capable of detecting this fluorochome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. Cytotoxicity-activating receptor that may contribute to the increased efficiency of activated natural killer (NK) cells to mediate tumor cell lysis.
Specifications

Specifications

CD335 (NKp46)
Mouse
IgG2a, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
29A1.4
PE-eFluor™ 610
Flow Cytometry
Primary
Monoclonal
PBS with 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.