missing translation for 'onlineSavingsMsg'
Learn More

CD335 (NKp46), eFluor 660, clone: 29A1.4, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-3351-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15311900

  • £82.86 / 25µg


EU Stock 1
Add to basket

Description

Description

Description: The monoclonal antibody 29A1.4 recognizes mouse NKp46, also known as CD335. CD335, a member of the natural cytotoxicity receptor (NCR) family, is a glycoprotein with 2 Ig-like domains and a short cytoplasmic tail. Expression of CD335 is uniquely found on NK cells (including immature NK cells, defined as DX5- CD3-, and thereby allowing discrimination between NKT cells and NK cells (NKp46+, CD3-). Furthermore, unlike many of the NK markers which also stain NKT cells, staining with 29A1.4 is not strain specific. Staining has been shown on C57Bl/6, SJL, CBA/CA and BALB/C strains. NKp46 has been shown to play a role in NK cell-mediated lysis of several tumor cells and pathogen-infected cell lines.The 29A1.4 monoclonal antibody has been shown to activate NK cells in vitro. The 29A1.4 monoclonal antibody does not deplete NK cells in vivo.Applications Reported: This 29A1.4 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 29A1.4 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD335 (NKp46)
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q8C567
CD335, LY94, NK-p46, NKP46, NK cell-activating receptor, lymphocyte antigen 94 homolog activating NK-receptor, NK-p46, natural killer cell p46-related protein
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
29A1.4
eFluor 660
Ncr1
Conjugated
Ncr1
IgG2a, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
17086
Videos

For Research Use Only.