missing translation for 'onlineSavingsMsg'
Learn More

CD335 (NKp46), PerCP-eFluor™ 710, clone: 9E2, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-3359-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15519246

  • £218.92 / 100 tests


Estimated Shipment date
from Supplier 03-05-2018
Add to basket

Description

Description

Description: This 9E2 monoclonal antibody reacts with human CD335, which is also known as NKp46. Expressed on NK cells, CD335 is a member of the natural cytotoxicity receptor family that also includes NKp30 and NKp44. This type I transmembrane glycoprotein is involved in activating and expanding resting NK cells following interaction with dendritic cells to eliminate tumor and virus-infected (e.g., influenza) cells. CD335 also interacts with pancreatic beta cells during the development of type I diabetes.The 9E2 antibody has been reported to induce cytotoxicity.Applications Reported: This 9E2 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 9E2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD335 (NKp46)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
O76036
CD335, LY94, NK-p46, NKP46, NK cell-activating receptor, lymphocyte antigen 94 homolog activating NK-receptor, NK-p46, natural killer cell p46-related protein
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
9.00E+02
PerCP-eFluor 710
NCR1
Conjugated
NCR1
IgG1, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
9437
Videos

For Research Use Only.