missing translation for 'onlineSavingsMsg'
Learn More

CD335 (NKp46), Functional Grade, clone: 9E2, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 16-3359-85

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15557786

  • £250.06 / 500Āµg


EU Stock 1
Add to basket

Description

Description

Description: This 9E2 monoclonal antibody reacts with human CD335, which is also known as NKp46. Expressed on NK cells, CD335 is a member of the natural cytotoxicity receptor family that also includes NKp30 and NKp44. This type I transmembrane glycoprotein is involved in activating and expanding resting NK cells following interaction with dendritic cells to eliminate tumor and virus-infected (e.g., influenza) cells. CD335 also interacts with pancreatic beta cells during the development of type I diabetes.The 9E2 antibody has been reported to induce cytotoxicity.Applications Reported: This 9E2 antibody has been reported for use in flow cytometric analysis and to induce cytotoxicity in functional assays.Applications Tested: This 9E2 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Endotoxin: Less than 0.001 ng/ug antibody as determined by the LAL assay.Storage and handling: Use in a sterile environment.Filtration: 0.2μm post-manufacturing filtered.Purity: Greater than 90%, as determined by SDS-PAGE.Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay.Aggregation: Less than 10%, as determined by HPLC.
Specifications

Specifications

CD335 (NKp46)
Monoclonal
1mg/mL
PBS with no preservative; pH 7.2
O76036
CD335, LY94, NK-p46, NKP46, NK cell-activating receptor, lymphocyte antigen 94 homolog activating NK-receptor, NK-p46, natural killer cell p46-related protein
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow CytometryĆ¾Functional Assay
9.00E+02
Functional Grade
NCR1
Purified
NCR1
IgG1, kappa
500μg
Store at 2-8°C.
Primary
9437
Videos

For Research Use Only.