missing translation for 'onlineSavingsMsg'
Learn More

CD33 Mouse anti-Human, PerCP-Cyanine5.5, Clone: WM-53 (WM53), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 45-0338-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15569016

  • £115.00 / 25 tests

Estimated Shipment
29-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD33 is a transmembrane protein of the sialic acid-binding immunoglobulin-like lectin (Siglec) family. It belongs to the immunoreceptor tyrosine-based inhibitory motif (ITIM)-containing molecules able of recruiting protein tyrosine phosphatases SHP-1 and SHP-2 to signal assemblies, and these ITIMs are also used for ubiquitin-mediated removal of the receptor from the cell surface. CD33 is expressed on cells of myelomonocytic lineage, binds sialic acid residues in N- and O-glycans on cell surfaces, and is a therapeutic target for acute myeloid leukemia. Further, CD33 is found on granulocyte and macrophage precursors in the bone marrow, but is not on pluripotent stem cells. CD33 is also expressed on, and is a useful marker for, peripheral monocytes. CD33 is useful for distinguishing myelogenous leukemia cells from lymphoid or erythroid leukemias. Diseases associated with CD43 dysfunction include gallbladder lymphoma and extracutaneous mastocytoma.
Specifications

Specifications

CD33
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P20138
FLJ00391, SIGLEC-3, p67
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
WM-53 (WM53)
PerCP-Cyanine5.5
CD33
Liquid
CD33
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
945
Videos
Documents

For Research Use Only.