missing translation for 'onlineSavingsMsg'
Learn More

CD33, APC, clone: WM-53 (WM53), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-0338-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15557916

  • £168.34 / 100 tests


EU Stock 1
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The WM-53 monoclonal antibody reacts with human CD33, also known as GP67 and P67, a 67kDa type I transmembrane glycoprotein that is a member of the Siglec (sialic acid-binding Ig superfamily lectin) family. It is highly specific to the hematopoietic compartment and is expressed on monocytes, activated T cells, granulocytes, myeloid progenitors, and mast cells.Applications Reported: This WM-53 (WM53) antibody has been reported for use in flow cytometric analysis.Applications Tested: This WM-53 (WM53) antibody has been pre-titrated and tested by flow cytometric analysis or normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD33
WM-53 (WM53)
APC
Conjugated
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.125μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Sialic Acid-Binding Immunoglobulin-Like Lectin 3, SIGLEC3
IgG1
RUO
Primary
Human
Videos