missing translation for 'onlineSavingsMsg'
Learn More

CD326 (EpCAM) Rat anti-Mouse, APC-eFluor® 780, Clone: G8.8, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 47-5791-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15589586

  • £203.00 / 100µg

In stock, ready to ship: 2

Please to purchase this item. Need a web account? Register with us today!


Description

Description

APC-eFluor 780 emits at 780 nm and is excited with the Red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Light sensitivity: This tandem is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL cell sample + 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. Ep-CAM is a ∽40 kD glycoprotein, which functions as an epithelial cell adhesion molecule. Ep-CAM mediates Ca2+-independent homotypic interactions. Formation of Ep-CAM-mediated adhesions has a negative regulatory effect on adhesions mediated by classic cadherins, which may have strong effects on the differentiation and growth of epithelial cells. Ep-CAM overexpression was suggested to be associated with enhanced epithelial proliferation. Ep-CAM is expressed in the majority of human carcinomas and is a marker for tumors of epithelial lineage.
Specifications

Specifications

CD326 (EpCAM)
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q99JW5
Epithelial Specific Antigen, ESA, KSA, M4S1, MK-1, DIAR5, EGP-2, EGP40, KS1/4, MIC18, TROP1, TACSTD1, EPCAM, Epithelial cell adhesion molecule
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
G8.8
APC-eFluor™ 780
Epcam
Liquid
Epcam
IgG2a, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
17075
Videos
Documents

For Research Use Only.