missing translation for 'onlineSavingsMsg'
Learn More

CD320 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA567296

Additional Details : Weight : 0.01000kg

Product Code. 15919964

  • £410.00 / 100µL
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: ASPLSTPTSAQ AAGPSSGSCP PTKFQCRTSG LCVPLTWRCD RDLDCSDGSD Highest antigen sequence identity to the following orthologs - mouse 59%, rat 60%.

Germinal center-B (GC-B) cells differentiate into memory B-cells and plasma cells (PC) through interaction with T-cells and follicular dendritic cells (FDC). CD320 augments the proliferation of PC precursors generated by IL-10. Receptor for the cellular uptake of transcobalamin bound cobalamin.
Specifications

Specifications

CD320
Polyclonal
Unconjugated
CD320
8D6A; CD320; TCblR; 8D6 antigen; 8D6AFDC-SM-8D6; CD320 antigenFDC-signaling molecule 8D6; 8D6TCBLR; CD320 molecule; TCblR; Transcobalamin receptor
Rabbit
Antigen Affinity Chromatography
RUO
51293
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunofluorescence
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9NPF0
CD320
Recombit Human CD320
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only