missing translation for 'onlineSavingsMsg'
Learn More

CD32 Rat, PE, Clone: 2.4G2, enQuireBio™

Rat Monoclonal Antibody

Brand:  EnquireBio QAB87-PE-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15962588

  • £219.00 / 100µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD32
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
FCGR2b
Protein A/G Purified
RUO
14131
IgG2b
Flow Cytometry
2.4G2
PE
XP_006496722
Rat
100 μg
Primary
Store at 2-4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers