missing translation for 'onlineSavingsMsg'
Learn More

CD32, eFluor 450, clone: 6C4 (CD32), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-0329-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15599656

  • £186.79 / 100 tests


EU Stock 5
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This 6C4 monoclonal antibody reacts with human CD32 (also known as Fc gammaRII). This antibody recognizes two isoforms of the receptor, Fc gammaRIIA and Fc gammaRIIB. These 40-kDa polymorphic transmembrane glycoproteins are expressed on B cells, granulocytes, monocytes, macrophages, and platelets. Moreover, these receptors are detected on natural killer cells. CD32 enables interaction between Fc gammaRII-expressing cells and opsonized antigen or IgG-containing immune complexes. Both receptors exhibit low affinity towards IgG and play a role in inflammation and autoimmune disease. This clone has been reported to inhibit Ig binding in a rosette blocking assay.Applications Reported: This 6C4 (CD32) antibody has been reported for use in flow cytometric analysis.Applications Tested: This 6C4 (CD32) antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD32
6C4 (CD32)
eFluor 450
Conjugated
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Low Affinity IgG Receptor 2a, FcGR2a, IGFR2
IgG1
RUO
Primary
Human
Videos