missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD32 Mouse anti-Human, eFluor 450, Clone: 6C4 (CD32), eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 48-0329-41
Code : NEW
Additional Details : Weight : 0.23750kg
Description
This 6C4 monoclonal antibody reacts with human CD32 (also known as Fc gammaRII). This antibody recognizes two isoforms of the receptor, Fc gammaRIIA and Fc gammaRIIB. These 40kDa polymorphic transmembrane glycoproteins are expressed on B cells, granulocytes, monocytes, macrophages, and platelets. Moreover, these receptors are detected on natural killer cells. CD32 enables interaction between Fc gammaRII-expressing cells and opsonized antigen or IgG-containing immune complexes. Both receptors exhibit low affinity towards IgG and play a role in inflammation and autoimmune disease. This clone has been reported to inhibit Ig binding in a rosette blocking assay.Specifications
CD32 | |
Monoclonal | |
5μL (0.25μg)/test | |
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer | |
Mouse | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
6C4 (CD32) | |
eFluor 450 | |
Low Affinity IgG Receptor 2a, FcGR2a, IGFR2 | |
25 Tests | |
Primary | |
Store at 2-8°C. Do not freeze. Light-sensitive material. |
For Research Use Only.