Learn More
CD319 (CRACC) Monoclonal Antibody (162), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 46-2229-42
173.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.23750kg
Description
SLAMF7 contains one Ig-like C2-type (immunoglobulin-like) domain. Isoform 1 mediates NK cell activation through a SAP-independent extracellular signal-regulated ERK-mediated pathway. It may play a role in lymphocyte adhesion. Isoform 3 does not mediate any activation. SAP can bind the cytoplasmic tail of isoform 1 when phosphorylated in the presence of Fyn (in vitro). SLAMF7 is expressed in spleen, lymph node, peripheral blood leukocytes, bone marrow, small intestine, stomach, appendix, lung and trachea. Expression was detected in NK cells, activated B-cells, NK-cell line but not in promyelocytic, B-, or T-cell lines. The isoform 3 is expressed at much lower level than isoform 1. There are three named isoforms.
CD319 (CRACC) Monoclonal antibody specifically detects CD319 (CRACC) in Human samples. It is validated for Flow Cytometry.Specifications
CD319 (CRACC) | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Q9NQ25 | |
SLAMF7 | |
Affinity chromatography | |
RUO | |
57823 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
162 | |
PerCP-eFluor 710 | |
SLAMF7 | |
19A; 19A24; 19A24 protein; 4930560D03Rik; CD2 subset 1; CD2-like receptor activating cytotoxic cells; CD2-like receptor-activating cytotoxic cells; CD319; CRACC; CS1; Leukocyte cell-surface antigen; membrane protein FOAP-12; Novel Ly9; novel LY9 (lymphocyte antigen 9) like protein; protein 19A; RGD1560190; SLAM family member 7; SLAMF7; UNQ576/PRO1138 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
For Research Use Only.