missing translation for 'onlineSavingsMsg'
Learn More

CD317 (BST2, PDCA-1), PerCP-eFluor™ 710, clone: eBio927, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-3172-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15311310

  • £70.58 / 25µg


EU Stock 3
Add to basket

Description

Description

Description: The eBio927 monoclonal antibody reacts with PDCA-1 (BST2, CD317), a specific marker of plasmacytoid dendritic cells (pDC), also known as type I IFN-producing cells (IPC) in the naive mouse. Mouse IPCs are typically CD11c+, CD11b-, B220+, Ly-6C+, and CD62L+. PDCA-1 is predominantly expressed by IPCs in the naive mouse which represents a very minor population (<0.5%) of splenocytes. It is upregulated on numerous cell types following stimulation which triggers an IFN response. PDCA-1 cycles between cell surface and intracellular compartments and may function to regulate trafficking of secreted cytokines. PDCA-1 (BST2) is the protein recognized by antibody 120G8. The eBio927 monoclonal antibody has also been shown to have functional activity. The epitope recognized by eBio927 is distinct from eBio129c; thus, the antibodies can be used to costain, purify and identify pDCs.Applications Reported: This eBio927 antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBio927 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.PerCP-eFluor® 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative. Our testing indicates that PerCP-eFluor® 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4°C, but please evaluate for alternative fixation protocols.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD317
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q8R2Q8
Bone marrow stromal cell antigen 2, Tetherin, PDCA-1, 2310015I10Rik, Bst-2, C87040, DAMP-1, GREG, DAMP-1 protein homolog, HM1.24 antigen, bone marrow stromal antigen 2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
eBio927
PerCP-eFluor 710
Bst2
Conjugated
Bst2
IgG2b, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
69550
Videos

For Research Use Only.