missing translation for 'onlineSavingsMsg'
Learn More

CD317 (BST2, PDCA-1), PE-eFluor 610, clone: eBio927, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 61-3172-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15540487

  • £68.96 / 25µg


Estimated Shipment date
from Supplier 28-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The eBio927 monoclonal antibody reacts with PDCA-1 (BST2, CD317), a specific marker of plasmacytoid dendritic cells (pDC), also known as type I IFN-producing cells (IPC) in the naive mouse. Mouse IPCs are typically CD11c+, CD11b-, B220+, Ly-6C+, and CD62L+. PDCA-1 is predominantly expressed by IPCs in the naive mouse which represents a very minor population (<0.5%) of splenocytes. It is upregulated on numerous cell types following stimulation which triggers an IFN response. PDCA-1 cycles between cell surface and intracellular compartments and may function to regulate trafficking of secreted cytokines. PDCA-1 (BST2) is the protein recognized by antibody 120G8. The eBio927 monoclonal antibody has also been shown to have functional activity. The epitope recognized by eBio927 is distinct from eBio129c; thus, the antibodies can be used to costain, purify and identify pDCs.Applications Reported: This eBio927 antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBio927 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.PE-eFluor® 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red®). Please make sure that your instrument is capable of detecting this fluorochome.Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD317 (BST2/PDCA-1)
eBio927
PE-eFluor 610
Conjugated
Rat
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
plasmacytoid DC Ag-1
IgG2b
RUO
Primary
Murine
Videos