missing translation for 'onlineSavingsMsg'
Learn More

CD317 (BST2, PDCA-1) Rat anti-Mouse, APC, Clone: eBio129c (129c), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-3171-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15578076

  • £78.50 / 25µg

Estimated Shipment
02-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. BST2, also known as CD317, HM1.24 antigen, DAMP-2 and recently designated tetherin, is an integral membrane protein with a novel topology. The specific function of the BST2, encoded by the bone marrow stromal cell antigen 2 gene, is undetermined; however, it may play a role in pre-B-cell growth, rheumatoid arthritis, and in antiretroviral defense, that blocks release of retrovirus from the cell surface. BST2 is highly expressed during B-cell development, from pro-B precursors to plasma cells, T-cells, monocytes, NK cells and DCs (at protein level).
Specifications

Specifications

CD317 (BST2, PDCA-1)
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q8R2Q8
Bone marrow stromal cell antigen 2, Tetherin, PDCA-1, 2310015I10Rik, Bst-2, C87040, DAMP-1, GREG, DAMP-1 protein homolog, HM1.24 antigen, bone marrow stromal antigen 2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
eBio129c (129c)
APC
Bst2
Liquid
Bst2
IgG2b, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
69550
Videos
Documents

For Research Use Only.