missing translation for 'onlineSavingsMsg'
Learn More

CD317 (BST2, PDCA-1), Functional Grade, clone: 26F8, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 16-3179-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15228607

  • £75.29 / 25µg


Estimated Shipment
28-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This 26F8 monoclonal antibody reacts with human CD317 (also known as BST2 and tetherin). CD317 is a 30-36-kDa type II transmembrane protein expressed on B cells and bone marrow stromal cells. Although reports have indicated that CD317 mRNA is detectable in activated T cells, protein expression in primary T cells and macrophages is undetectable. Moreover, certain T cell lines, such as Jurkat, do not express detectable levels of CD317 protein. CD317 has been associated with pre-B cell growth and the terminal differentiation of plasma B cells. More recently, this molecule has been reported to prevent HIV-1 virion release from the surface of infected cells, leading to reuptake and degradation of the virus. This activity is inhibited by the HIV-1 accessory protein Vpu. CD317 has been identified as the ligand for the ILT7 receptor.Applications Reported: This 26F8 antibody has been reported for use in flow cytometric analysis and functional assays. The 26F8 clone has been reported to block the binding of CD317 to the ILT7 receptor.Applications Tested: This 26F8 antibody has been tested by flow cytometric analysis on MCF7 cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Endotoxin: Less than 0.001 ng/ug antibody as determined by the LAL assay.Storage and handling: Use in a sterile environment.Filtration: 0.2μm post-manufacturing filtered.Purity: Greater than 90%, as determined by SDS-PAGE.Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay.Aggregation: Less than 10%, as determined by HPLC.
Specifications

Specifications

CD317 (BST2/PDCA-1)
26F8
Unconjugated
Purified
IgG1
RUO
Primary
Human
Functional Assay
1mg/mL
aqueous buffer, no sodium azide
Murine
25μg
Store at 2-8°C.
Monoclonal
Videos