Learn More
CD314 (NKG2D) Monoclonal Antibody (CX5), Super Bright™ 600, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£137.00 - £313.00
Specifications
Antigen | CD314 (NKG2D) |
---|---|
Clone | CX5 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933564
|
Affymetrix eBioscience
63-5882-82 |
100 μg |
£313.00
100µg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923564
|
Invitrogen
63-5882-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The CX5 monoclonal antibody reacts with the mouse NKG2D, a lectin-like molecule expressed on both human and mouse NK cells. Mouse NKG2D binds to retinoic acid-inducible RAE-1 alpha, beta, gamma, delta, epsilon and the minor histocompatibility molecule H60 and has the ability to costimulate multiple NK activation receptors, through the DAP12/DAP10 adaptor molecules. NKG2D is expressed by all spleen and liver NK cells, NK1.1^+ thymocytes, in vitro activated LAK cells, and a subset of splenic NKT cells. Applications Reported: This CX5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CX5 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 1.0 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 600 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 600 nm. We recommend using a 610/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No.
CD314 (NKG2D) Monoclonal antibody specifically detects CD314 (NKG2D) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD314 (NKG2D) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
O54709 | |
Klrk1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Klrk1 |
CX5 | |
Flow Cytometry | |
Super Bright 600 | |
Rat | |
Mouse | |
27007 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |