missing translation for 'onlineSavingsMsg'
Learn More

CD314 (NKG2D), PerCP-eFluor™ 710, clone: CX5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-5882-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15559296

  • £223.65 / 100µg


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Description

Description

Description: The CX5 monoclonal antibody reacts with the mouse NKG2D, a lectin-like molecule expressed on both human and mouse NK cells. Mouse NKG2D binds to retinoic acid-inducible RAE-1 alpha, beta, gamma, delta, epsilon and the minor histocompatibility molecule H60 and has the ability to costimulate multiple NK activation receptors, through the DAP12/DAP10 adaptor molecules. NKG2D is expressed by all spleen and liver NK cells, NK1.1^+ thymocytes, in vitro activated LAK cells, and a subset of splenic NKT cells.Applications Reported: This CX5 antibody has been reported for use in flow cytometric analysis.Applications Tested: This CX5 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD314 (NKG2D)
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
O54709
CD314, NK cell receptor D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, killer cell lectin-like receptor subfamily K member 1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
CX5
PerCP-eFluor 710
Klrk1
Conjugated
Klrk1
IgG1, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Primary
27007
Videos

For Research Use Only.